Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha'-Dilaurin 96.0+%, TCI America™
SDP

Catalog No. G007910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G007910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G007910G Supplier TCI America Supplier No. G007910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 539-93-5
Molecular Formula C27H52O5
Molecular Weight (g/mol) 456.708
MDL Number MFCD00048423
InChI Key KUVAEMGNHJQSMH-UHFFFAOYSA-N
Synonym Glycerol alpha,alpha′C-Dilaurate
PubChem CID 101758
IUPAC Name (3-dodecanoyloxy-2-hydroxypropyl) dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)O

Specifications

Melting Point 59°C
Color White
Quantity 10 g
Formula Weight 456.71
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dilaurin
EINECSNumber (2)-0669&(2)-0771
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.