accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53914-95-7

IUPAC Name: 2,5,8,11,14,19,22,25,28,31-decaoxatricyclo[30.2.2.2¹⁵,¹⁸]octatriaconta-1(34),15,17,32,35,37-hexaene
Synonyms: 2,5,8,11,14,19,22,25,28,31-decaoxatricyclo[30.2.2.2¹⁵,¹⁸]octatriaconta-1(34),15,17,32,35,37-hexaene
Molecular Weight (g/mol): 536.62
Molecular Formula: C28H40O10
InChi Key: REKDBTBSNFSNGP-UHFFFAOYSA-N
SMILES: C1COCCOC2=CC=C(OCCOCCOCCOCCOC3=CC=C(OCCOCCO1)C=C3)C=C2
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results