accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53934-75-1

OONHNNH2NHOOOHO(E/Z)

CAS RN 53934-75-1

IUPAC Name: 5-{[amino({[(benzyloxy)carbonyl]amino})methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino({[(benzyloxy)carbonyl]amino})methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 442.47
Molecular Formula: C22H26N4O6
InChi Key: YWMQITGDXCEQSI-UHFFFAOYNA-N
SMILES: NC(NC(=O)OCC1=CC=CC=C1)=NCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results