Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha,Nomega-Dicarbobenzoxy-L-arginine 98.0+%, TCI America™
SDP

Catalog No. D11791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11791G Supplier TCI America Supplier No. D11791G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53934-75-1
Molecular Formula C22H26N4O6
Molecular Weight (g/mol) 442.472
MDL Number MFCD00070573
InChI Key YWMQITGDXCEQSI-SFHVURJKSA-N
Synonym Nalpha,Nomega-Di-Cbz-L-arginine, alpha,omega-Di-Z-Arg-OH
PubChem CID 44630235
IUPAC Name (2S)-5-[[amino(phenylmethoxycarbonylamino)methylidene]amino]-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)NC(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 442.47
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha,Nomega-Dicarbobenzoxy-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.