accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5394-23-0

ClNClN

CAS RN 5394-23-0

IUPAC Name: 4,7-dichloro-1,10-phenanthroline
Synonyms: 4,7-dichloro-1,10-phenanthroline
Molecular Weight (g/mol): 249.09
Molecular Formula: C12H6Cl2N2
InChi Key: GIEQBYJCGYHHSU-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC3=C(Cl)C=CN=C3C2=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results