accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CHC

CAS RN 5396-61-2

IUPAC Name: 3-ethylhept-1-yn-3-ol
Synonyms: 3-ethylhept-1-yn-3-ol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: TXCLTIFWSNGTIK-UHFFFAOYNA-N
SMILES: CCCCC(O)(CC)C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results