Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-1-heptyn-3-ol, TCI America™
SDP

Catalog No. E02725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02725ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02725ML Supplier TCI America Supplier No. E02725ML

Chemical Identifiers

CAS 5396-61-2
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00041677
InChI Key TXCLTIFWSNGTIK-UHFFFAOYSA-N
Synonym (1-Ethyl-1-hydroxypentyl)acetylene
PubChem CID 220863
IUPAC Name 3-ethylhept-1-yn-3-ol
SMILES CCCCC(CC)(C#C)O

Specifications

Color Yellow
Boiling Point 165°C
UN Number 1987
Quantity 5 mL
Formula Weight 140.23
Physical Form Liquid
Chemical Name or Material 3-Ethyl-1-heptyn-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.