accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53963-03-4

IUPAC Name: deca-1,5-diyne
Synonyms: deca-1,5-diyne
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: IWIDILFDSIIOSB-UHFFFAOYSA-N
SMILES: CCCCC#CCCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results