Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Decadiyne 97.0+%, TCI America™
SDP

Catalog No. D17245ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17245ML 5 mL
D17241ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17245ML Supplier TCI America Supplier No. D17245ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53963-03-4
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00041651
InChI Key IWIDILFDSIIOSB-UHFFFAOYSA-N
PubChem CID 104660
IUPAC Name deca-1,5-diyne
SMILES CCCCC#CCCC#C

Specifications

Color Yellow
Boiling Point 79°C
Quantity 5 mL
UN Number 3295
Formula Weight 134.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Decadiyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.