accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5398-34-5

IUPAC Name: sodium 8-aminonaphthalene-1,3,6-trisulfonic acid
Synonyms: sodium 8-aminonaphthalene-1,3,6-trisulfonic acid
Molecular Weight (g/mol): 406.35
Molecular Formula: C10H9NNaO9S3
InChi Key: AFFMXGGDMXALPG-UHFFFAOYSA-N
SMILES: [Na+].NC1=C2C(=CC(=C1)S(O)(=O)=O)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)