accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CHNNHOCH3H3CCH3CH3CH3H3COCH3CH3H3C(E/Z)(E/Z)

CAS RN 539834-19-0

IUPAC Name: 2,4-di-tert-butyl-6-{[(5-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}bicyclo[2.2.1]heptan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one
Synonyms: 2,4-di-tert-butyl-6-{[(5-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}bicyclo[2.2.1]heptan-2-yl)amino]methylidene}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 558.85
Molecular Formula: C37H54N2O2
InChi Key: SMGJFOOBGSAVJS-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=CC(=CNC2CC3CC2CC3NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results