Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R,4R,5R)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane 98.0+%, TCI America™
SDP

Catalog No. B2652100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2652100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2652100MG Supplier TCI America Supplier No. B2652100MG

Chemical Identifiers

CAS 539834-19-0
Molecular Formula C37H54N2O2
Molecular Weight (g/mol) 558.851
InChI Key SMGJFOOBGSAVJS-FXYBLIFYSA-N
Synonym 2,2′C-[(1R,2R,4R,5R)-Bicyclo[2.2.1]heptane-2,5-diylbis(nitrilomethyl)]bis(4,6-di-tert-butylphenol)
PubChem CID 133554169
IUPAC Name (6Z)-2,4-ditert-butyl-6-[[[2-[[(E)-(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]-5-bicyclo[2.2.1]heptanyl]amino]methylidene]cyclohexa-2,4-dien-1-one
SMILES CC(C)(C)C1=CC(=CNC2CC3CC2CC3NC=C4C=C(C=C(C4=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C

Specifications

Melting Point 252°C
Color Yellow
Quantity 100 mg
Formula Weight 558.85
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R,4R,5R)-2,5-Bis(3,5-di-tert-butyl-2-hydroxybenzylideneamino)bicyclo[2.2.1]heptane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.