CAS RN 5399-02-0
IUPAC Name:
heptadecanenitrile
Synonyms:
heptadecanenitrile
Molecular Weight (g/mol):
251.46
Molecular Formula:
C17H33N
InChi Key:
ZXPWFWWSCFIFII-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCC#N
1
–
1
of
1
results
1
Heptadecanonitrile 95.0+%, TCI America™
CAS: 5399-02-0 Molecular Formula: C17H33N Molecular Weight (g/mol): 251.458 MDL Number: MFCD00042635 InChI Key: ZXPWFWWSCFIFII-UHFFFAOYSA-N Synonym: Cetyl Cyanide, Hexadecyl Cyanide, Heptadecanenitrile PubChem CID: 79388 IUPAC Name: heptadecanenitrile SMILES: CCCCCCCCCCCCCCCCC#N