Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptadecanonitrile 95.0+%, TCI America™
SDP

Catalog No. H056825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H056825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H056825G Supplier TCI America Supplier No. H056825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5399-02-0
Molecular Formula C17H33N
Molecular Weight (g/mol) 251.458
MDL Number MFCD00042635
InChI Key ZXPWFWWSCFIFII-UHFFFAOYSA-N
Synonym Cetyl Cyanide, Hexadecyl Cyanide, Heptadecanenitrile
PubChem CID 79388
IUPAC Name heptadecanenitrile
SMILES CCCCCCCCCCCCCCCCC#N

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 208°C
Quantity 25 g
Formula Weight 251.46
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecanonitrile
EINECSNumber (2)-1507
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.