accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(R)CH3(R)CH3(R)H3COHH3CH3CO

CAS RN 54-28-4

IUPAC Name: (2R)-2,7,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Synonyms: gamma-tocopherol D-gamma-tocopherol gamma-tocopherol skeleton gamma-tocopherol, d-
Molecular Weight (g/mol): 416.69
Molecular Formula: C28H48O2
InChi Key: QUEDXNHFTDJVIY-DQCZWYHMSA-N
SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=C(C)C(C)=C2O1
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results