Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-γ-Tocopherol, MilliporeSigma™ Supelco™

Catalog No. 111014552 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4552 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4552 Supplier MilliporeSigma Supelco Supplier No. 47785
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 54-28-4
Physical Form Neat
Linear Formula C28H48O2
MDL Number MFCD00066529
Synonym (R,R,R)-gamma-Tocopherol; 7,8-Dimethyltocol
Type Food Additive Standard
Quantity 25 mg
Formula Weight 416.68
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.