accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54-47-7

IUPAC Name: [(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methoxy]phosphonic acid
Synonyms: pyridoxal phosphate pyridoxal 5-phosphate pyromijin pidopidon vitahexin-P vitahexin P apolon B(sub 6) hi-pyridoxin pyridoxal 5'-phosphate phosphate, pyridoxal
Molecular Weight (g/mol): 247.14
Molecular Formula: C8H10NO6P
InChi Key: NGVDGCNFYWLIFO-UHFFFAOYSA-N
SMILES: CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results