Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal 5-Phosphate (Monohydrate), Fisher BioReagents

Catalog No. BP26761 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP26761 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP26761

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • PLP is useful for modifying lysyl and valyl residues and functions as a coenzyme in transamination reactions by forming a Schiff-base linkage with lysine groups on aminotransferase
  • PLP is an active form of vitamin B7
  • Also serves as a coenzyme in some decarboxylation and deamination reactions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-47-7
Molecular Formula C8H10NO6P
Molecular Weight (g/mol) 247.14
MDL Number MFCD00006333
InChI Key NGVDGCNFYWLIFO-UHFFFAOYSA-N
Synonym pyridoxal phosphate, codecarboxylase, pyridoxal 5-phosphate, pyridoxal 5'-phosphate, pyridoxal p, biosechs, hairoxal, pyromijin, vitazechs, hiadelon
PubChem CID 1051
ChEBI CHEBI:18405
IUPAC Name (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate
SMILES CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O

Specifications

Melting Point 140°C
Color White
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98%
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 8092
Formula Weight 247.14
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material Pyridoxal 5-Phosphate, Monohydrate
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage Store below 0°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.