accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 540-49-8

IUPAC Name: (1Z)-1,2-dibromoethene
Synonyms: ethene, 1,2-dibromo-, (Z)-
Molecular Weight (g/mol): 185.85
Molecular Formula: C2H2Br2
InChi Key: UWTUEMKLYAGTNQ-UPHRSURJSA-N
SMILES: Br\C=C/Br
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)