accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeP(R)(R)P(R)(R)CH3CH3CH3CH3

CAS RN 540475-45-4

IUPAC Name: 1,1'-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ferrocene
Synonyms:
Molecular Weight (g/mol): 414.29
Molecular Formula: C22H32FeP2
InChi Key:
SMILES: [Fe].C[C@@H]1CC[C@@H](C)P1c1cccc1.C[C@@H]1CC[C@@H](C)P1c1cccc1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results