Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-Vapol Hydrogenphosphate

Catalog No. 675512100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-551-2100MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-551-2100MG Supplier Sigma Aldrich Supplier No. 675512100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful chiral Bronsted acid catalyst in the addition of sulfonamides to Boc-activated aryl imines.

Specifications

CAS 540475-45-4
Melting Point 99.9°C to 101.5°C
Boiling Point 35°C to 36°C
Molecular Formula C22H32FeP2
Linear Formula C22H32FeP2
Quantity 500 mg
Synonym R,R-Me-Ferrocelane(TM)
Molecular Weight (g/mol) 414.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.