accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COHO

CAS RN 5409-31-4

IUPAC Name: 3,4-diethoxybenzoic acid
Synonyms: BENZOICACID34diethoxy
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: VVKCVAPLTRZJHH-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1OCC)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)