Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D408025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D408025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D408025G Supplier TCI America Supplier No. D408025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5409-31-4
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00002504
InChI Key VVKCVAPLTRZJHH-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-diethoxy, benzoicacid34diethoxy, 3,4-diethoxy-benzoic acid, acmc-1apyn, ksc271i5p, 3,4-bis ethyloxy benzoic acid, 3,4-diethoxybenzoic acid
PubChem CID 79417
IUPAC Name 3,4-diethoxybenzoic acid
SMILES CCOC1=C(C=C(C=C1)C(=O)O)OCC

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 25 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.