accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5409-37-0

IUPAC Name: 6-amino-N,N,7-trimethylacridin-3-aminium chloride
Synonyms: 6-amino-N,N,7-trimethylacridin-3-aminium chloride
Molecular Weight (g/mol): 287.79
Molecular Formula: C16H18ClN3
InChi Key: JRMDFAKCPRMZKA-UHFFFAOYSA-N
SMILES: [Cl-].C[NH+](C)C1=CC2=NC3=CC(N)=C(C)C=C3C=C2C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results