Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coriphosphine O, TCI America™
SDP

Catalog No. C05391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C05391G Supplier TCI America Supplier No. C05391G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5409-37-0
Molecular Formula C16H18ClN3
Molecular Weight (g/mol) 287.79
MDL Number MFCD00050595
InChI Key JRMDFAKCPRMZKA-UHFFFAOYSA-N
Synonym Basic Yellow 7
PubChem CID 9860722
IUPAC Name 6-amino-N,N,7-trimethylacridin-3-aminium chloride
SMILES [Cl-].C[NH+](C)C1=CC2=NC3=CC(N)=C(C)C=C3C=C2C=C1

Specifications

Quantity 1 g
Formula Weight 287.79
Physical Form Crystalline Powder
Chemical Name or Material Coriphosphine O
RTECSNumber AR7343600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.