accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CCH3HOOO

CAS RN 541-15-1

IUPAC Name: 3-hydroxy-4-(trimethylazaniumyl)butanoate
Synonyms: carnitine 卡尼汀
Molecular Weight (g/mol): 161.2
Molecular Formula: C7H15NO3
InChi Key: PHIQHXFUZVPYII-UHFFFAOYNA-N
SMILES: C[N+](C)(C)CC(O)CC([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (11)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Grade 
  • (1)
  • (6)
  • (5)
Physical Form 
  • (2)
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (11)
  • (1)
  • (11)
  • (2)
  • (6)
  • (6)
  • (8)
  • (6)
  • (5)
  • (2)
  • (3)
  • (11)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (5)
  • (2)
  • (3)
  • (5)