Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levocarnitine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014545 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4545 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4545 Supplier MilliporeSigma Supelco Supplier No. PHR15691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Levocarnitine is a naturally available compound that plays a significant role in fatty acid oxidation and energy production in the human body. It is majorly found in skeletal and cardiac muscles of mammals and facilitates the transport of long chain fatty acids into mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

CAS 541-15-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula (CH3)3 N+CH2 CH(OH)CH2 COO-
MDL Number MFCD00038747
Melting Point 197°C to 212°C
Synonym L-Carnitine inner salt; (-)-(R)-3-Hydroxy-4-(trimethylammonio)butyrate; Vitamin BT
Type Certified Reference Material
Quantity 1 g
Formula Weight 161.20
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.