accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 541-57-1

IUPAC Name: 4-hydroxy-2,5-dihydrofuran-2-one
Synonyms: 4-hydroxy-5H-furan-2-one
Molecular Weight (g/mol): 100.07
Molecular Formula: C4H4O3
InChi Key: JZQBAGOECGRTSA-UHFFFAOYSA-N
SMILES: OC1=CC(=O)OC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results