accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 541-70-8

IUPAC Name: benzene-1,3-diamine; sulfuric acid
Synonyms: developer C; sulfuric acid
Molecular Weight (g/mol): 206.22
Molecular Formula: C6H10N2O4S
InChi Key: LDXYDHGRKFMULJ-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC(N)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results