Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Phenylenediamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. P017425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P017425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P017425G Supplier TCI America Supplier No. P017425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 541-70-8
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.216
MDL Number MFCD00060223
InChI Key LDXYDHGRKFMULJ-UHFFFAOYSA-N
Synonym 1,3-Diaminobenzene Sulfate
PubChem CID 10942
IUPAC Name benzene-1,3-diamine;sulfuric acid
SMILES C1=CC(=CC(=C1)N)N.OS(=O)(=O)O

Specifications

Color Purple-White
Quantity 25 g
UN Number 2811
Formula Weight 206.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylenediamine Sulfate
EINECSNumber (3)-0185
RTECSNumber SS7700005
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.