accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH3

CAS RN 541-85-5

IUPAC Name: 5-methylheptan-3-one
Synonyms: methylheptanone
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: PSBKJPTZCVYXSD-UHFFFAOYNA-N
SMILES: CCC(C)CC(=O)CC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)