Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-3-heptanone 96%, Thermo Scientific™

Catalog No. AC297080010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC297080010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC297080010 Supplier Thermo Scientific Chemicals Supplier No. 297080010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-85-5
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00009340
InChI Key PSBKJPTZCVYXSD-UHFFFAOYNA-N
Synonym 5-methyl-3-heptanone, 3-heptanone, 5-methyl, ethyl sec-amyl ketone, ethyl 2-methylbutyl ketone, 3-methyl-5-heptanone, ethyl amylketone, 2-methylbutyl ethyl ketone, 5-methyl-3-heptanon, ethyl amyl cetone, 5-methyl 3-heptanone
PubChem CID 7822
IUPAC Name 5-methylheptan-3-one
SMILES CCC(C)CC(=O)CC

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Density 0.82
Infrared Spectrum Authentic
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Linear Formula CH3CH2COCH2CH(CH3)CH2CH3
Quantity 1 L
Beilstein 01, I, 363
Merck Index 15, 6156
Formula Weight 128.21
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 5-Methyl-3-heptanone, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.