accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5410-29-7

IUPAC Name: (2-nitrophenyl)arsonic acid
Synonyms: arsonic acid, (2-nitrophenyl)-
Molecular Weight (g/mol): 247.04
Molecular Formula: C6H6AsNO5
InChi Key: UYEDGVZDGVIURN-UHFFFAOYSA-N
SMILES: O[As](O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results