Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenylarsonic Acid 98.0+%, TCI America™
SDP

Catalog No. N013325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N013325G 25 g
N01331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N013325G Supplier TCI America Supplier No. N013325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5410-29-7
Molecular Formula C6H6AsNO5
Molecular Weight (g/mol) 247.038
MDL Number MFCD00047664
InChI Key UYEDGVZDGVIURN-UHFFFAOYSA-N
Synonym 2-nitrobenzenearsonic acid, arsonic acid, 2-nitrophenyl, o-nitrophenylarsenic acid, o-nitrobenzenearsonic acid, o-nitrophenyl arsonic acid, benzenearsonic acid, o-nitro, unii-hnr3su2y5p, 2-nitrophenyl arsonic acid, arsonic acid, as-2-nitrophenyl, hnr3su2y5p
PubChem CID 79424
IUPAC Name (2-nitrophenyl)arsonic acid
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])[As](=O)(O)O

Specifications

Quantity 25 g
UN Number 3465
Formula Weight 247.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenylarsonic Acid
RTECSNumber CY5950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.