CAS RN 5411-12-1
IUPAC Name:
2-benzoyl-3-phenyloxirane
Synonyms:
2-benzoyl-3-phenyloxirane
Molecular Weight (g/mol):
224.26
Molecular Formula:
C15H12O2
InChi Key:
UQGMJZQVDNZRKT-UHFFFAOYNA-N
SMILES:
O=C(C1OC1C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
1,3-Diphenyl-2,3-epoxy-1-propanone 98.0+%, TCI America™
CAS: 5411-12-1 Molecular Formula: C15H12O2 Molecular Weight (g/mol): 224.26 MDL Number: MFCD00022336 InChI Key: UQGMJZQVDNZRKT-UHFFFAOYNA-N Synonym: 2-benzoyl-3-phenyloxirane,chalcone oxide,1,3-diphenyl-2,3-epoxy-1-propanone,chalcone epoxide,trans-chalcone oxide,phenyl 3-phenyloxiran-2-yl methanone,ccris 2214,trans-phenyl 3-phenyloxiranyl methanone,methanone, phenyl 3-phenyloxiranyl,phenyl 3-phenyl-2-oxiranyl methanone PubChem CID: 92219 IUPAC Name: phenyl-(3-phenyloxiran-2-yl)methanone SMILES: C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3