Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diphenyl-2,3-epoxy-1-propanone 98.0+%, TCI America™
SDP

Catalog No. D43785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43785G 5 g
D43781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43785G Supplier TCI America Supplier No. D43785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5411-12-1
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00022336
InChI Key UQGMJZQVDNZRKT-UHFFFAOYNA-N
Synonym 2-benzoyl-3-phenyloxirane, chalcone oxide, 1,3-diphenyl-2,3-epoxy-1-propanone, chalcone epoxide, trans-chalcone oxide, phenyl 3-phenyloxiran-2-yl methanone, ccris 2214, trans-phenyl 3-phenyloxiranyl methanone, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone
PubChem CID 92219
IUPAC Name phenyl-(3-phenyloxiran-2-yl)methanone
SMILES C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 90°C
Color White
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenyl-2,3-epoxy-1-propanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.