accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54127-30-9

IUPAC Name: (5,6-dichloropyridin-3-yl)methanol
Synonyms: (5,6-dichloropyridin-3-yl)methanol
Molecular Weight (g/mol): 178.01
Molecular Formula: C6H5Cl2NO
InChi Key: ZOFUUOULXZPZHP-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results