Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichloro-3-pyridinemethanol 96.0+%, TCI America™
SDP

Catalog No. D24025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24025G 5 g
D24021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24025G Supplier TCI America Supplier No. D24025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54127-30-9
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.012
MDL Number MFCD00671515
InChI Key ZOFUUOULXZPZHP-UHFFFAOYSA-N
Synonym 2,3-Dichloro-5-hydroxymethylpyridine, 5,6-Dichloronicotinyl Alcohol
PubChem CID 21882961
IUPAC Name (5,6-dichloropyridin-3-yl)methanol
SMILES C1=C(C=NC(=C1Cl)Cl)CO

Specifications

Melting Point 76°C
Quantity 5 g
Formula Weight 178.01
Percent Purity ≥96.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloro-3-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.