CAS RN 5413-65-0
IUPAC Name:
1,2,4,5,8-pentahydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms:
1,2,4,5,8-pentahydroxyanthracene-9,10-dione
Molecular Weight (g/mol):
288.21
Molecular Formula:
C14H8O7
InChi Key:
LLDSAQJCCUZHBA-UHFFFAOYSA-N
SMILES:
OC1=CC(O)=C2C(=O)C3=C(O)C=CC(O)=C3C(=O)C2=C1O
1
–
1
of
1
results
1
MP Biomedicals, Inc Alizarin Cyanin, MP Biomedicals
CAS: 5413-65-0 Molecular Formula: C14H8O7 Molecular Weight (g/mol): 288.211 InChI Key: LLDSAQJCCUZHBA-UHFFFAOYSA-N PubChem CID: 407615 IUPAC Name: 1,2,4,5,8-pentahydroxyanthracene-9,10-dione SMILES: C1=CC(=C2C(=C1O)C(=O)C3=C(C2=O)C(=C(C=C3O)O)O)O