Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alizarin Cyanin, MP Biomedicals

Catalog No. ICN206439 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206439 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206439 Supplier MP Biomedicals, Inc Supplier No. 0520643910

Chemical Identifiers

CAS 5413-65-0
Molecular Formula C14H8O7
Molecular Weight (g/mol) 288.211
InChI Key LLDSAQJCCUZHBA-UHFFFAOYSA-N
PubChem CID 407615
IUPAC Name 1,2,4,5,8-pentahydroxyanthracene-9,10-dione
SMILES C1=CC(=C2C(=C1O)C(=O)C3=C(C2=O)C(=C(C=C3O)O)O)O

Specifications

Formula Weight 358.19
Quantity 10 g
Chemical Name or Material Alizarin Cyanin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.