CAS RN 54135-60-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Apoptosis Inhibitor
The Apoptosis Inhibitor, also referenced under CAS 54135-60-3, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.