Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Apoptosis Inhibitor

Catalog No. 17848810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-848-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-848-810MG Supplier MilliporeSigma™ Supplier No. 17848810MG
Add to Cart
Add to Cart

The Apoptosis Inhibitor, also referenced under CAS 54135-60-3, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins apoptosis induction
CAS 54135-60-3
Molecular Formula C13H16O4
Formula Weight 236.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from moisture
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.