accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOClClOH2ONa

CAS RN 5415-26-9

IUPAC Name: sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]-2-methylbenzen-1-olate hydrate
Synonyms: sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]-2-methylbenzenolate hydrate
Molecular Weight (g/mol): 322.12
Molecular Formula: C13H10Cl2NNaO3
InChi Key: IWXAXMKDZIDRGC-UHFFFAOYSA-M
SMILES: O.[Na+].CC1=C([O-])C=CC(=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results