Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3'-methylindophenol Sodium Salt Hydrate [Redox Indicator], TCI America™
SDP

Catalog No. D03791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D03791G Supplier TCI America Supplier No. D03791G

Chemical Identifiers

CAS 5415-26-9
Molecular Formula C13H10Cl2NNaO3
Molecular Weight (g/mol) 322.117
MDL Number MFCD00019446
InChI Key IWXAXMKDZIDRGC-UHFFFAOYSA-M
Synonym 2,6-Dichlorophenol-indo-o-cresol Sodium Salt
PubChem CID 131697927
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]-2-methylphenolate;hydrate
SMILES CC1=C(C=CC(=C1)N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].O.[Na+]

Specifications

Color Black-Purple
Quantity 1 g
Formula Weight 304.10
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-3′-methylindophenol Sodium Salt Hydrate [Redox Indicator]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.