accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 541542-62-5

IUPAC Name: N-butyl-4-chlorobutanamide
Synonyms: N-butyl-4-chlorobutanamide
Molecular Weight (g/mol): 177.67
Molecular Formula: C8H16ClNO
InChi Key: MATMHSSBGIRYDI-UHFFFAOYSA-N
SMILES: CCCCNC(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results