accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5418-43-9

IUPAC Name: sodium 5-methyl-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]-2-(propan-2-yl)benzen-1-olate
Synonyms: sodium 2-isopropyl-5-methyl-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzenolate
Molecular Weight (g/mol): 277.3
Molecular Formula: C16H16NNaO2
InChi Key: VSHCTHOZOJWONF-UHFFFAOYSA-M
SMILES: [Na+].CC(C)C1=CC(N=C2C=CC(=O)C=C2)=C(C)C=C1[O-]
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results