Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Isopropyl-2'-methylindophenol Sodium Salt [Redox Indicator], TCI America™
SDP

Catalog No. M04081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M04081G Supplier TCI America Supplier No. M04081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5418-43-9
Molecular Formula C16H16NNaO2
Molecular Weight (g/mol) 277.299
MDL Number MFCD00058962
InChI Key VSHCTHOZOJWONF-UHFFFAOYSA-M
Synonym Thymolindophenol Sodium Salt
PubChem CID 44630378
IUPAC Name sodium;5-methyl-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]-2-propan-2-ylphenolate
SMILES CC1=C(C=C(C(=C1)[O-])C(C)C)N=C2C=CC(=O)C=C2.[Na+]

Specifications

Quantity 1 g
Formula Weight 277.30
Physical Form Crystalline Powder
Chemical Name or Material 5′-Isopropyl-2′-methylindophenol Sodium Salt [Redox Indicator]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.