CAS RN 54187-96-1
IUPAC Name:
1-(chloromethyl)-1H-1,2,3-benzotriazole
Synonyms:
1-(chloromethyl)-1,2,3-benzotriazole
Molecular Weight (g/mol):
167.6
Molecular Formula:
C7H6ClN3
InChi Key:
VSEROABGEVRIRY-UHFFFAOYSA-N
SMILES:
ClCN1N=NC2=CC=CC=C12
1
–
1
of
1
results
1
1-(Chloromethyl)-1H-benzotriazole 98.0+%, TCI America™
CAS: 54187-96-1 Molecular Formula: C7H6ClN3 Molecular Weight (g/mol): 167.60 MDL Number: MFCD00216649 InChI Key: VSEROABGEVRIRY-UHFFFAOYSA-N PubChem CID: 3328222 IUPAC Name: 1-(chloromethyl)-1H-1,2,3-benzotriazole SMILES: ClCN1N=NC2=CC=CC=C12