Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Chloromethyl)-1H-benzotriazole 98.0+%, TCI America™
SDP

Catalog No. C15265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15265G 5 g
C152625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15265G Supplier TCI America Supplier No. C15265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54187-96-1
Molecular Formula C7H6ClN3
Molecular Weight (g/mol) 167.60
MDL Number MFCD00216649
InChI Key VSEROABGEVRIRY-UHFFFAOYSA-N
PubChem CID 3328222
IUPAC Name 1-(chloromethyl)-1H-1,2,3-benzotriazole
SMILES ClCN1N=NC2=CC=CC=C12

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 167.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Chloromethyl)-1H-benzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.