accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-02-9

CH3NNH2NH2NN

CAS RN 542-02-9

IUPAC Name: 6-methyl-1,3,5-triazine-2,4-diamine
Synonyms: acetoguanamine
Molecular Weight (g/mol): 125.14
Molecular Formula: C4H7N5
InChi Key: NJYZCEFQAIUHSD-UHFFFAOYSA-N
SMILES: CC1=NC(N)=NC(N)=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)